Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Betacellulin/BTC Antibody (RM0154-9H1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Betacellulin/BTC |
---|---|
Clone | RM0154-9H1 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Description
Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Betacellulin/BTC | |
LYOPH | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rat | |
Cell Cycle and Replication, Signal Transduction | |
Q05928 | |
685 | |
IgG2 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
RM0154-9H1 | |
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
betacellulin, probetacellulin | |
BTC | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title