Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Betacellulin/BTC Antibody (RM0154-9H1), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212053IR
Description
Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Betacellulin/BTC | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BTC | |
Mouse recombinant Betacellulin/BTC | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0154-9H1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
betacellulin, probetacellulin | |
Rat | |
Protein A or G purified | |
Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction | |
685 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction