Learn More
Invitrogen™ betacellulin Monoclonal Antibody (34362)

Description
In Western blots, this antibody does not cross-react with recombinant human (rh) EGF, rhHB-EGF, rhTGF- alpha, rhHRG- alpha, or rhAR under non-reducing conditions. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | betacellulin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 34362 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | BTC |
| Gene Accession No. | P35070 |
| Gene Alias | Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.