Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ betacellulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547044
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547044 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547044 Supplier Invitrogen™ Supplier No. PA547044
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human BTC is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen betacellulin
Applications Immunohistochemistry, Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene BTC
Gene Accession No. Q05928
Gene Alias Bcn; betacellulin; betacellulin, epidermal growth factor family member; BTC; OTTHUMP00000160600; OTTHUMP00000219057; Probetacellulin
Gene Symbols BTC
Host Species Goat
Immunogen E. coli-derived recombinant mouse Betacellulin/BTC Asp32-Gln118.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12223
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.