Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betahistine Hydrochloride, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18600932 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-932 100 g Amber Glass Bottle
18-600-930 5 g Amber Glass Bottle
18-600-931 25 g Amber Glass Bottle
18-600-933 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-932 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B1976100GM
Add to Cart
Add to Cart
Request a Quote

B1976, 5579-84-0, C8H12N2·2HCl

Spectrum™ Chemical Betahistine Hydrochloride, USP is an anti-vertigo drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5579-84-0
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.11
InChI Key XVDFMHARQUBJRE-UHFFFAOYSA-N
IUPAC Name dihydrogen methyl[2-(pyridin-2-yl)ethyl]amine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CNCCC1=CC=CC=N1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 99 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.