Betaine hydrochloride, 99%

Manufacturer: Alfa Aesar A1612230

Quantity 250g

 View more versions of this product

Catalog No. AAA1612230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 590-46-5
    Molecular Formula (CH3)3NCH2CO2·HCl
    Formula Weight 153.61
    MDL Number MFCD00011903
    Beilstein 3916181
    Merck Index 14,1179
    Synonym (Carboxymethyl)trimethylammonium chloride
    Chemical Name or Material Betaine hydrochloride

    Specifications

    Formula Weight 153.61
    Quantity 250g
    TSCA TSCA
    Molecular Formula (CH3)3NCH2CO2·HCl
    Melting Point ≈240°C (dec.)
    MDL Number MFCD00011903
    CAS 590-46-5
    Beilstein 3916181
    Merck Index 14,1179
    Synonym (Carboxymethyl)trimethylammonium chloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.
    RTECS Number BP3136000
    Chemical Name or Material Betaine hydrochloride
    Assay 99%

    Description

    Applications
    Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

    Solubility
    Soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit