Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone Dipropionate, Micronized Powder, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18600948 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

Catalog No. 18-600-948

Supplier: Spectrum Chemical Manufacturing Corporation BE18125GM

Only null left
Add to Cart
Add to Cart

BE181, 5593-20-4, C28H37FO7

Spectrum™ Chemical Betamethasone Dipropionate, Micronized Powder, USP is a glucocorticoid steroid applied as a topical cream, ointment, lotion or gel to treat itching and other minor skin conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5593-20-4
Molecular Formula C28H37FO7
Molecular Weight (g/mol) 504.60
InChI Key CIWBQSYVNNPZIQ-XYWKZLDCSA-N
IUPAC Name 2-[(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
SMILES CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

Specifications

Packaging Amber Glass Bottle
CAS Max % 100%
Industry Type Pharmaceutical
Quantity 25 g
Percent Purity 97 to 103%
Grade USP
Chemical Name or Material Betamethasone Dipropionate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.