Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone Sodium Phosphate, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18600953 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail

BE183, 151-73-5, C22H28FNa2O8P

Spectrum™ Chemical Betamethasone Sodium Phosphate, USP is a strong glucocorticoid steroid used to reduce inflammation, irritation, and pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 151-73-5
Molecular Formula C22H28FNa2O8P
Molecular Weight (g/mol) 516.41
InChI Key PLCQGRYPOISRTQ-LWCNAHDDSA-L
IUPAC Name disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation +99° to +105°
Percent Purity 97 to 103%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.