Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ betatrophin Monoclonal Antibody (OTI1C6)
GREENER_CHOICE

Catalog No. PIMA527083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527083 Supplier Invitrogen™ Supplier No. MA527083
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

betatrophin Monoclonal Antibody for Western Blot, IHC (P)

The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen betatrophin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANGPTL8
Gene Accession No. Q6UXH0
Gene Alias angiopoietin like 8; angiopoietin-like 8; angiopoietin-like protein 8; Angptl8; betatrophin; betatrophin variant 1; betatrophin variant 2; C19orf80; EG624219; Gm6484; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; hepatocellular carcinoma-associated protein TD26 homolog; lipasin; LOC100361444; predicted gene 6484; PRO1185; PVPA599; rCG31799-like; refeeding-induced fat and liver protein; Rifl; TD26; UNQ599/PRO1185
Gene Symbols ANGPTL8
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 22-198 of C19ORF80 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55908
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.