Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ betatrophin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538043
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538043 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538043 Supplier Invitrogen™ Supplier No. PA538043
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is rat liver tissue lysate. At least three isoforms of Betatrophin are known to exist; this antibody will detect the two largest isoforms.

The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen betatrophin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ANGPTL8
Gene Accession No. Q6UXH0, Q8R1L8
Gene Alias angiopoietin like 8; angiopoietin-like 8; angiopoietin-like protein 8; Angptl8; betatrophin; betatrophin variant 1; betatrophin variant 2; C19orf80; EG624219; Gm6484; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; hepatocellular carcinoma-associated protein TD26 homolog; lipasin; LOC100361444; predicted gene 6484; PRO1185; PVPA599; rCG31799-like; refeeding-induced fat and liver protein; Rifl; TD26; UNQ599/PRO1185
Gene Symbols ANGPTL8
Host Species Rabbit
Immunogen 16 amino acid peptide near the amino terminus of human Betatrophin.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100361444, 55908, 624219
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.