Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaxolol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7123565
Click to view available options
Quantity:
50 mg
250 mg

Chemical Identifiers

CAS 63659-19-8
Molecular Formula C18H30ClNO3
Molecular Weight (g/mol) 343.892
MDL Number MFCD00242959
InChI Key CHDPSNLJFOQTRK-UHFFFAOYSA-N
Synonym betaxolol hydrochloride, betaxolol hcl, betoptima, kerlone, betoptic, kerlon, lokren, betaxolo hydrochloride, betoptic s
PubChem CID 107952
ChEBI CHEBI:643228
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 50 mg
Formula Weight 343.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Betaxolol Hydrochloride
RTECSNumber UA9823000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.