Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaxolol hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Catalog No. p-4783281
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective β1 antagonist

Selective β1-adrenoceptor antagonist.

Chemical Identifiers

CAS 63659-19-8
Molecular Formula C18H30ClNO3
Molecular Weight (g/mol) 343.892
InChI Key CHDPSNLJFOQTRK-UHFFFAOYSA-N
Synonym betaxolol hydrochloride, betaxolol hcl, betoptima, kerlone, betoptic, kerlon, lokren, betaxolo hydrochloride, betoptic s
PubChem CID 107952
ChEBI CHEBI:643228
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl

Specifications

Quantity 10 mg
Formula Weight 343.89
Percent Purity >99%
Chemical Name or Material Betaxolol hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.