Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bethanechol Chloride, 100.2%

Catalog No. ICN19016025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19016025 25 g
ICN19016005 5 g
ICN19016080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19016025 Supplier MP Biomedicals, Inc Supplier No. 0219016025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 590-63-6
Molecular Formula C7H17ClN2O2
Molecular Weight (g/mol) 196.675
InChI Key XXRMYXBSBOVVBH-UHFFFAOYSA-N
Synonym bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone
PubChem CID 11548
ChEBI CHEBI:3085
IUPAC Name 2-carbamoyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)N.[Cl-]

Specifications

Color White
Assay Percent Range 100.2%
Quantity 25 g
Formula Weight 196.7
Physical Form Powder
Chemical Name or Material Bethanechol Chloride
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.