Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Betula pendula BETVIA Chimeric Recombinant Rabbit Monoclonal Antibody (M0418)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548032
Description
Specificity: This antibody binds to Bet v 1, in particular to the naturally occurring isoforms Bet v 1.0101 and Bet v 1.0102, but not to Bet v 1.0112.
May be a general steroid carrier protein.
Specifications
Betula pendula BETVIA Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Rabbit | |
Protein A | |
RUO | |
Plant | |
Antibody | |
IgG λ |
ELISA, In vitro Assay | |
M0418 | |
Unconjugated | |
allergen Bet v 1-A; Allergen Bet v I-A; BETVI; Major pollen allergen Bet v 1-A | |
Bet v 1-A, a major allergen of the European white birch. | |
200 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction