Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Betulinic acid
GREENER_CHOICESDP

Catalog No. 390610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-061-0 10 mg
39-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-061-0 Supplier Tocris Bioscience™ Supplier No. 3906/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antitumor and anti-HIV agent; activates NF-κB. Also TGR5 agonist

Betulinic acid is a natural triterpenoid that displays anti-HIV and antitumor activity. Induces the production of reactive oxygen species (ROS) and activates NF-κB. Also a GPBA receptor partial agonist (EC50 = 1.04 μM, efficacy 83%). Betulinic acid suppresses areobic glycolysis by regulating the Cav-1/NF-κB/c-Myc pathway in and inhibits growth in breast cancer cells.

Chemical Identifiers

CAS 472-15-1
Molecular Formula C30H48O3

Specifications

Quantity 10 mg
Formula Weight 456.7
Purity >98%
Molecular Weight (g/mol) 456.711
Chemical Name or Material Betulinic acid
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.