Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BEX1/BEX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100404 Supplier Invitrogen™ Supplier No. PA5100404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BEX1/2.

BEX1/BEX2 belongs to the brain expressed X-linked gene family. The encoded protein interacts with the transcription factor LIM domain only 2 in a DNA-binding complex that recognizes the E-box element and promotes transcription. BEX1/BEX2 has been found to be a tumor suppressor that is silenced in human glioma. In breast cancer cells, this gene product modulates apoptosis in response to estrogen and tamoxifen, and enhances the anti-proliferative effect of tamoxifen. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BEX1/BEX2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BEX1
Gene Accession No. Q3MKQ1, Q3MKQ2, Q9BXY8, Q9HBH7, Q9R224, Q9WTZ8
Gene Alias AA407839; AL024066; AU018692; BEX1; BEX2; Bex4; brain expressed gene 1; brain expressed X-linked 1; brain expressed X-linked 2; brain expressed, X-linked 1; brain-expressed X-linked protein 1; Brain-expressed X-linked protein 1 homolog; brain-expressed X-linked protein 2; Brain-expressed X-linked protein 2 homolog; DJ79P11.1; EG2RVC; expression gene 2 in rat visual cortex; hBex2; HGR74-h; ovarian granulosa cell 13.0 kDa protein hGR74; protein BEX1; Protein BEX2; protein hBex2; reduced expression 3; reduced expression protein 3; Rex3; REX-3
Gene Symbols BEX1, BEX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BEX2(Accession Q9BXY8), corresponding to amino acid residues M1-R50.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12069, 19716, 363498, 501625, 55859, 84707
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.