Learn More
Invitrogen™ BEX1 Polyclonal Antibody

Description
This antibody is predicted to react with non-human primate based on sequence homology.
Specifications
Specifications
| Antigen | BEX1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | BEX1 |
| Gene Accession No. | Q9HBH7 |
| Gene Alias | AA407839; AU018692; BEX1; BEX2; Bex4; brain expressed gene 1; brain expressed X-linked 1; brain expressed, X-linked 1; brain-expressed X-linked protein 1; Brain-expressed X-linked protein 1 homolog; EG2RVC; H HGR74-h; HBEX2; HGR74-h; ovarian granulosa cell 13.0 kDa protein hGR74; protein BEX1; reduced expression 3; reduced expression protein 3; Rex3; REX-3 |
| Gene Symbols | BEX1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.