Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BEX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560925 Supplier Invitrogen™ Supplier No. PA560925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WAIPNRHIEH NEARDDVERF VGQMMEIKRK TREQQMRHYM RFQTPEPDNH YDF Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 72%.

Bex4 may play a role in microtubule deacetylation by negatively regulating the SIRT2 deacetylase activity toward alpha-tubulin and thereby participate to the control of cell cycle progression and genomic stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BEX4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Bex4
Gene Accession No. Q9NWD9
Gene Alias BEX family member 4; BEX1-like 1; BEX1-like protein 1; Bex4; BEXL1; brain expressed gene 4; brain expressed X-linked 4; brain expressed X-linked-like 1; brain expressed, X-linked 4; Brain-expressed X-linked protein 4; brain-expressed X-linked protein 4 homolog; FLJ10097; NADE3; Nerve growth factor receptor-associated protein 3; protein BEX4
Gene Symbols Bex4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BEX4. Recombinant protein control fragment (Product #RP-100369).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.