Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BEX4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS7978R

Description
Bex4 may play a role in microtubule deacetylation by negatively regulating the SIRT2 deacetylase activity toward alpha-tubulin and thereby participate to the control of cell cycle progression and genomic stability.Specifications
BEX4 | |
Polyclonal | |
Unconjugated | |
Bex4 | |
BEX family member 4; BEX1-like protein 1; BEX4; BEXL1; brain expressed X-linked 4; brain expressed X-linked-like 1; brain expressed, X-linked 4; Brain-expressed X-linked protein 4; FLJ10097; NADE3; Nerve growth factor receptor-associated protein 3; Protein BEX4 | |
Rabbit | |
Protein A | |
RUO | |
56271 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NWD9 | |
Bex4 | |
KLH conjugated synthetic peptide derived from human BEX4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction