Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bexarotene, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J63701MCPM
Click to view available options
Quantity:
100 mg

Synthetic retinoid analog with specific affinity for the retinoid X receptor

Applications

Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer's model lowered the most toxic form of B-amyloid peptide and increased cognitive ability.

Solubility

Soluble in DMSO and ethanol

Notes

Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153559-49-0
Molecular Formula C24H28O2
Molecular Weight (g/mol) 348.49 g/mol
MDL Number MFCD00932428
InChI Key NAVMQTYZDKMPEU-UHFFFAOYSA-N
Synonym bexarotene, targretin, targrexin, targretyn, targret, bexarotenum, bexarotene usan, bexaroteno, bexaroten, targretin-gel
PubChem CID 82146
ChEBI CHEBI:50859
IUPAC Name 4-[1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethenyl]benzoic acid
SMILES CC1=CC2=C(C=C1C(=C)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC2(C)C

Specifications

Melting Point 220°C to 225°C
Color White
Quantity 100 mg
Merck Index 14,1194
Solubility Information Soluble in DMSO and ethanol
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Bexarotene
RTECSNumber DH6834830
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.