Learn More
Bexarotene, 98%, Thermo Scientific Chemicals
Description
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer's model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bexarotene is used as a Synthetic retinoid analog with specific affinity for the retinoid X receptor, an antineoplastic agent, already approved as an oral antineoplastic agent for cutaneous T cell lymphoma and being investigated against other cancers. A study has found that bexarotene in a mouse Alzheimer′s model lowered the most toxic form of β-amyloid peptide and increased cognitive ability.
Solubility
Soluble in DMSO and ethanol
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 220°C to 225°C |
| Color | White |
| Quantity | 300 mg |
| Merck Index | 14,1194 |
| Solubility Information | Soluble in DMSO and ethanol |
| Formula Weight | 348.48 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Chemical Name or Material | Bexarotene |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| RTECSNumber | DH6834830 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.