Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BF 2649 hydrochloride
GREENER_CHOICE

Catalog No. 374310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-431-0 10 mg
37-435-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-431-0 Supplier Tocris Bioscience™ Supplier No. 3743/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

H3 receptor inverse agonist/antagonist

Potent and selective histamine H3 receptor inverse agonist (EC50 = 1.5 nM, Ki = 0.16 nM). Exhibits nootropic effects in cognitive disorders; reduces locomotor hyperactivity induced by methamphetamine. Brain penetrant.

Chemical Identifiers

CAS 903576-44-3
Molecular Formula C17H27Cl2NO

Specifications

Quantity 10 mg
Formula Weight 332.31
Purity >99%
Molecular Weight (g/mol) 332.309
Chemical Name or Material BF 2649 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.