Learn More
Specifications
Specifications
| Gene ID (Entrez) | 51283 |
| Species | Human |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56093 |
| Gene Alias | BARRNF47RING finger protein 47, bifunctional apoptosis inhibitor, bifunctional apoptosis regulator |
| Gene Symbol | BFAR |
| Quantity | 0.1 mg |
| Research Category | Apoptosis, Zinc Finger |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
