Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BFAR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $304.50
Specifications
Antigen | BFAR |
---|---|
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056093T
![]() |
Novus Biologicals
NB10056093SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB10056093
![]() |
Novus Biologicals
NB10056093 |
0.05 mL |
Each for $304.50
|
|
|||||
Description
BFAR Polyclonal specifically detects BFAR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
BFAR | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Zinc Finger | |
Whole antisera with 0.05% Sodium Azide | |
51283 | |
A synthetic peptide corresponding to amino acids 3-21 (EPQKSYVNTMDLERDEPLK) of human BAR was used as immunogen; GenBank no. NP_057645.1. Human BAR is a 450 amino acid protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
BARRNF47RING finger protein 47, bifunctional apoptosis inhibitor, bifunctional apoptosis regulator | |
BFAR | |
IgG | |
Unpurified | |
BAR |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title