Learn More
BFSP1 Mouse anti-Human, Clone: OTI3H6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804079

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
BFSP1 | |
Monoclonal | |
Unconjugated | |
BFSP1 | |
CP115, CP94, CTRCT33, LIFL-H | |
Mouse | |
Affinity Chromatography | |
RUO | |
631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3H6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q12934 | |
BFSP1 | |
Human recombit protein fragment corresponding to amino acids 405-665 of human BFSP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.