Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BFSP2 Monoclonal Antibody (OTI1C2)
GREENER_CHOICE

Catalog No. PIMA526663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526663 Supplier Invitrogen™ Supplier No. MA526663
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BFSP2 Monoclonal Antibody for Western Blot, IHC (P)

More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, the protein product of this gene (phakinin), and filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene have been associated with juvenile-onset, progressive cataracts and Dowling-Meara epidermolysis bullosa simplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BFSP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BFSP2
Gene Accession No. Q13515
Gene Alias 49 kDa cytoskeletal protein; AI448593; beaded filament protein CP49; beaded filament structural protein 2; beaded filament structural protein 2, phakinin; bfps2, Cytoskeletal protein, 49-kD; BFSP2; CP 47; CP47; CP49; CTRCT12; DKEYP-73D8.2; hypothetical protein LOC494090; lens fiber cell beaded filament protein CP 47; Lens fiber cell beaded filament protein CP 49; lens intermediate filament-like light; LIFL-L; MGC142078; MGC142080; phakinin; PHAKOSIN; zgc:103750
Gene Symbols BFSP2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 184-415 of BFSP2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8419
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.