Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ BG-11 Media, optimized for cyanobacteria

Catalog No. A1379902
Encompass_Preferred
Click to view available options
Quantity:
1 L
6 L

BG-11 Media, optimized for cyanobacteria

Gibco™ BG-11 Media is optimized for the growth and maintenance of select Cyanobacteria including Synechococcus elongates. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.

Each Gibco™ BG-11 Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helps you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Cyanobacteria culture, results in better overall efficiency and more robust data (Fig. 1, 2).

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1X
Content And Storage Gibco™ BG-11 Growth Media contains:

•  6 L Gibco™ BG-11 Media — Store at 4 °C
Form Liquid
Media Type Algal Media
Preparation Method None (Ready-to-Use)
Product Type Microbiology Media
Target Organism Class Cyanobacteria
pH 6 to 8
Quantity 6 L
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.