Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHA, FCC, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18600972
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle

BH104, 25013-16-5, C11H16O2

Spectrum™ Chemical BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.25
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES COC1=CC=C(O)C(=C1)C(C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Packaging Amber Glass Bottle
Quantity 12 kg
Percent Purity 98.5%
Grade FCC
Physical Form Crystals or Flakes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.