Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHC80/PHF21A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981378
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1378 Supplier Bioss Supplier No. BS11452R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHF21A (PHD finger protein 21A) is a component of the BHC complex, a corepressor complex that represses transcription of neuron-specific genes in non-neuronal cells. The BHC complex is recruited at RE1/NRSE sites by REST and acts by deacetylating and demethylating specific sites on histones, thereby acting as a chromatin modifier. In the BHC complex, PHF21A may act as a scaffold.

Specifications

Antigen BHC80/PHF21A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Phf21a
Gene Accession No. Q96BD5
Gene Alias 80kDa; Bhc; Bhc80; BHC80a; BM-006; Braf35/HDAC complex (Bhc); BRAF35/HDAC2 complex; BRAF35/HDAC2 complex (80 kDa); BRAF35-HDAC complex protein BHC80; D030065N23Rik; KIAA1696; LOC100345216; mBHC80; Pftf1; PHD finger protein 21A; Phf21a; RGD1560612
Gene Symbols Phf21a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PHF21A .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51317
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.