Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHE23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556510
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556510 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556510 Supplier Invitrogen™ Supplier No. PA556510
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLVAFLNQGQ GLAAPVNAAP LTPFGQATVC PFSAGAALGP CPDKCAAFSG TPSALCKHCH EKP Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes a member of the basic helix-loop-helix transcription factor family. Members of this family contain two highly conserved and functionally distinct domains: the basic domain targets sequence-specific DNA binding, while the helix-loop-helix domain facilitates protein interaction. Studies of a related gene in mouse suggest that the encoded protein may function as a transcriptional repressor in the pancreas and brain, and that it is required for normal retinal function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHE23
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Bhlhe23
Gene Accession No. Q8NDY6
Gene Alias A930001L02Rik; bA305P22.3; basic helix-loop-helix domain containing, class B, 4; basic helix-loop-helix domain containing, class B4; basic helix-loop-helix family member e23; basic helix-loop-helix family, member e23; BETA4; beta-cell E-box transactivating factor 4; BHLHB4; BHLHE23; Class B basic helix-loop-helix protein 4; class E basic helix-loop-helix protein 23; H20; RGD1559760
Gene Symbols Bhlhe23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BHE23. Recombinant protein control fragment (Product #RP-95507).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128408
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.