Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHLHE41 Polyclonal Antibody
BHLHE41 Polyclonal Antibody
Supplier: Thermo Scientific PA599229
Description
BHLHE41 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
BHLHE41 | |
Polyclonal | |
Unconjugated | |
Bhlhe41 | |
2-Dec; 6430520M22Rik; basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix domain containing, class B3; basic helix-loop-helix family member e41; basic helix-loop-helix family, member e41; bHLH transcriptional factor Dec2; Bhlhb2l; Bhlhb3; bHLHe41; Class B basic helix-loop-helix protein 3; class E basic helix-loop-helix protein 41; Dec2; differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; mDEC2; SHARP1; SHARP-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
117095, 79362, 79365 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q99PV5, Q9C0J9 | |
Bhlhe41 | |
A synthesized peptide derived from human BHLHE41(Accession Q9C0J9), corresponding to amino acid residues R11-N61. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction