Learn More
Invitrogen™ BHLHE41 Polyclonal Antibody

Description
Antibody detects endogenous levels of total BHLHB3.
Specifications
Specifications
| Antigen | BHLHE41 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Bhlhe41 |
| Gene Accession No. | Q99PV5, Q9C0J9 |
| Gene Alias | 2-Dec; 6430520M22Rik; basic helix-loop-helix domain containing, class B, 3; basic helix-loop-helix domain containing, class B3; basic helix-loop-helix family member e41; basic helix-loop-helix family, member e41; bHLH transcriptional factor Dec2; Bhlhb2l; Bhlhb3; bHLHe41; Class B basic helix-loop-helix protein 3; class E basic helix-loop-helix protein 41; Dec2; differentially expressed in chondrocytes protein 2; Enhancer-of-split and hairy-related protein 1; hDEC2; mDEC2; SHARP1; SHARP-1 |
| Gene Symbols | Bhlhe41 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.