Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHLHA9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237997
Description
BHLHA9 Polyclonal specifically detects BHLHA9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BHLHA9 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q7RTU4 | |
BHLHA9 | |
This antibody was developed against a recombinant protein corresponding to amino acids: SKARRMAANVRERKRILDYNEAFNALRRALRHDLGGKRLSKIATLRRAIHRIAALSLVLRASPAPRGPCGHLECHG | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Basic Helix-Loop-Helix Family, Member A9, bHLHa9, BHLHF42, Class A Basic Helix-Loop-Helix Protein 9, Class F Basic Helix-Loop-Helix Factor 42, Class II Basic Helix-Loop-Helix Protein | |
Rabbit | |
Affinity Purified | |
RUO | |
727857 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction