Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHLHA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564042 Supplier Invitrogen™ Supplier No. PA564042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKARRMAANV RERKRILDYN EAFNALRRAL RHDLGGKRLS KIATLRRAIH RIAALSLVLR ASPAPRGPCG HLECHG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Putative transcription factor, which may play a role in limb development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHA9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BHLHA9
Gene Accession No. Q7RTU4
Gene Alias A830053O21Rik; basic helix-loop-helix family member a9; basic helix-loop-helix family, member a9; bHLHa9; bHLHf42; class A basic helix-loop-helix protein 9; Class B basic helix-loop-helix factor 42; class F basic helix-loop-helix factor 42; class II basic helix-loop-helix protein; Fingerin; novel helix-loop-helix DNA-binding domain containing protein; Orphan1a; wu:fb99e06
Gene Symbols BHLHA9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BHLHA9. Recombinant protein control fragment (Product #RP-103460).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727857
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.