Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLHA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568907 Supplier Thermo Scientific Supplier No. PA568907
Add to Cart
Add to Cart

BHLHA9 Polyclonal Antibody

BHLHA9 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHA9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BHLHA9
Gene Accession No. Q5RJB0
Gene Alias A830053O21Rik; basic helix-loop-helix family member a9; basic helix-loop-helix family, member a9; bHLHa9; bHLHf42; class A basic helix-loop-helix protein 9; Class B basic helix-loop-helix factor 42; class F basic helix-loop-helix factor 42; class II basic helix-loop-helix protein; Fingerin; novel helix-loop-helix DNA-binding domain containing protein; Orphan1a; wu:fb99e06
Gene Symbols BHLHA9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse BHLHA9.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 320522
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.