Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHLHB5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23903925UL
Description
BHLHB5 Polyclonal specifically detects BHLHB5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
BHLHB5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Q8NFJ8 | |
BHLHE22 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EDDLFLHKSLSASTSKRLEAAFRSTPPGMDLSLAPPPRERPASSSSSPLGCFEPADPEGAGLLLPPP | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Basic Helix-Loop-Helix Domain Containing, Class B, 5, basic helix-loop-helix family, member e22, Beta3, BHLHE22, CAGL85, Class B Basic Helix-Loop-Helix Protein 5, Class E Basic Helix-Loop-Helix Protein 22, TNRC20, Trinucleotide Repeat Containing 20, Trinucleotide Repeat-Containing Gene 20 Protein | |
Rabbit | |
Affinity Purified | |
RUO | |
27319 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction