Learn More
Invitrogen™ BHLHB5 Polyclonal Antibody

Description
Immunogen sequence: AAAAALHPAL GAYEQAAGYP FSAGLPPAAS CPEKCALFNS VSSSLCKQCT E Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | BHLHB5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BHLHE22 |
| Gene Accession No. | Q8NFJ8 |
| Gene Alias | basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family member e22; basic helix-loop-helix family, member e22; Beta3; BHLHB5; bHLHe22; CAGL85; Class B basic helix-loop-helix protein 5; class E basic helix-loop-helix protein 22; Protein BETA3; TNRC20; trinucleotide repeat containing 20; trinucleotide repeat-containing gene 20 protein |
| Gene Symbols | BHLHE22 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.