Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHLHB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100405 Supplier Invitrogen™ Supplier No. PA5100405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BHLHE22.

BHLHB5 inhibits DNA binding of TCF3/E47 homodimers and TCF3 (E47)/NEUROD1 heterodimers and acts as a strong repressor of Neurod1 and Myod-responsive genes, probably by heterodimerization with class a basic helix-loop-helix factors. BHLHB5 despite the presence of an intact basic domain, does not bind to DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHB5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BHLHE22
Gene Accession No. Q8C6A8, Q8NFJ8
Gene Alias basic helix-loop-helix (bHLH) gene, class B, beta3; basic helix-loop-helix domain containing, class B, 5; basic helix-loop-helix domain containing, class B5; basic helix-loop-helix family member e22; basic helix-loop-helix family, member e22; Beta3; BHLHB5; bHLHe22; CAGL85; Class B basic helix-loop-helix protein 5; class E basic helix-loop-helix protein 22; Protein BETA3; TNRC20; trinucleotide repeat containing 20; trinucleotide repeat-containing gene 20 protein
Gene Symbols BHLHE22
Host Species Rabbit
Immunogen A synthesized peptide derived from human BHLHE22(Accession Q8NFJ8), corresponding to amino acid residues L119-A169.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27319, 365748, 59058
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.