Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLHB9/p60TRP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981555
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1555 Supplier Bioss Supplier No. BS11653R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in the control of cellular aging and survival.

Specifications

Antigen BHLHB9/p60TRP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BHLHB9
Gene Accession No. Q6PI77
Gene Alias basic helix-loop-helix domain containing, class B, 9; BHLHB9; GASP3; KIAA1701; p60-like protein; p60TRP; Protein BHLHb9; Transcription regulator of 60 kDa
Gene Symbols BHLHB9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BHLHB9/p60TRP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80823
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.