Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHLHB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109250 Supplier Invitrogen™ Supplier No. PA5109250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: U937, A549, SH-SY-5Y, mouse brain tissue.

May play a role in the control of cellular aging and survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHB9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene BHLHB9
Gene Accession No. Q6PI77
Gene Alias 2700087I09Rik; basic helix-loop-helix domain containing, class B, 9; basic helix-loop-helix domain containing, class B9; BHLHB9; GASP3; KIAA1701; KKPROT; p60-like protein; p60TRP; Protein BHLHb9; transcription regulator of 60 kDa
Gene Symbols BHLHB9
Host Species Rabbit
Immunogen Synthetic peptide within Human BHLHB9 aa 498-547.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80823
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.