Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHLHE40 Monoclonal Antibody (OTI10H1)
GREENER_CHOICE

Catalog No. PIMA527272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527272 Supplier Invitrogen™ Supplier No. MA527272
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BHLHE40 Monoclonal Antibody for Western Blot, IHC (P)

Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHE40
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BHLHE40
Gene Accession No. O14503
Gene Alias 1-Dec; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; bHLHb2; Bhlhe40; C130042M06Rik; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; cytokine response gene 8; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of split and hairy-related protein 2; enhancer-of-split and hairy-related protein 2; FLJ99214; HLHB2; SHARP2; SHARP-2; stimulated by retinoic acid 14; stimulated by retinoic acid gene 13 protein; STRA13; Stra14
Gene Symbols BHLHE40
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-313 of BHLHE40 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8553
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.