Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRA13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597288 Supplier Invitrogen™ Supplier No. PA597288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEGAGAGSGF RKELVSRLLH LHFKDDKTKE AAVRGVRQAQ AEDALRVDVD QLEKVLPQLL LDF; Positive Samples: H460; Cellular Location: Chromosome, Nucleus, centromere, kinetochore.

STRA13 is a DNA-binding component of the FA core complex involved in DNA damage repair and genome maintenance.STRA13 is recruited to forks stalled by DNA interstrand cross-links, and required for cellular resistance to such lesions. As a component of the APITD1/CENPS complex, STRA13 is also essential for the stable assembly of the outer kinetchore.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRA13
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CENPX
Gene Accession No. A8MT69
Gene Alias CENPX; CENP-X; Centromere protein X; D9; FAAP10; FANCM-associated histone fold protein 2; FANCM-interacting histone fold protein 2; Fanconi anemia-associated polypeptide of 10 kDa; MGC14480; MHF2; retinoic acid-inducible gene D9 protein homolog; stimulated by retinoic acid 13; stimulated by retinoic acid 13 homolog; stimulated by retinoic acid gene 13 protein homolog; STRA13
Gene Symbols CENPX
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201254
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.