Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHLHE40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 178951AP150UL
Description
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Specifications
| BHLHE40 | |
| Polyclonal | |
| Unconjugated | |
| BHLHE40 | |
| BHLHB2, BHLHE40, DEC1, SHARP 2, SHARP2, STRA13, Stra14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20893, 79431, 8553 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14503, O35185, O35780 | |
| BHLHE40 | |
| BHLHE40 Fusion Protein Ag12236 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction