Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLHE41 Monoclonal Antibody (OTI10B11), TrueMAB™, OriGene
SDP

Catalog No. 501680981
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-0981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-0981 Supplier OriGene Supplier No. TA806146
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.

Specifications

Antigen BHLHE41
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene BHLHE41
Gene Accession No. Q9C0J9
Gene Alias BHLHB3, DEC2, SHARP1, hDEC2
Gene Symbols BHLHE41
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-297 of human BHLHE41 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79365
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.