Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLHE41 Mouse anti-Human, Clone: OTI10B11, lyophilized, TrueMAB™
SDP

Catalog No. 501667706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-7706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-7706 Supplier OriGene Supplier No. CF806146
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.

Specifications

Antigen BHLHE41
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10B11
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene BHLHE41
Gene Accession No. Q9C0J9
Gene Alias BHLHB3, DEC2, SHARP1, hDEC2
Gene Symbols BHLHE41
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-297 of human BHLHE41 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79365
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.