Learn More
BHLHE41 Mouse anti-Human, Clone: OTI7B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806408

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.Specifications
BHLHE41 | |
Monoclonal | |
Unconjugated | |
BHLHE41 | |
BHLHB3, DEC2, SHARP1, hDEC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79365 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9C0J9 | |
BHLHE41 | |
Human recombit protein fragment corresponding to amino acids 1-297 of human BHLHE41 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.