Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BHMG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569751
Description
This target displays homology in the following species: Human: 100%.
BHMG1 gene ontology annotations related to this gene include DNA binding; protein dimerization activity.
Specifications
BHMG1 | |
Polyclonal | |
Unconjugated | |
BHMG1 | |
basic helix-loop-helix and HMG box domain containing 1; basic helix-loop-helix and HMG box domain-containing protein 1; basic helix-loop-helix and HMG-box containing 1; BHMG1; HMG domain containing; HMGDC | |
Rabbit | |
Affinity chromatography | |
RUO | |
388553 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
C9JSJ3 | |
BHMG1 | |
synthetic peptide directed towards the N terminal region of human BHMG1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction