Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHMT Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP100161
Description
BHMT Polyclonal specifically detects BHMT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
BHMT | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5 | |
Goat | |
Affinity Purified | |
RUO | |
635 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, ELISA | |
0.5 mg/mL | |
Western Blot 0.03 - 0.1 ug/mL, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:16000 | |
NP_001704.1 | |
BHMT | |
Peptide with sequence C-EQQLKELFEKQK corresponding to C-Terminus according to NP_001704.1 . | |
0.1 mg | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction