Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHMT Antibody (OTI3E11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270250H
Description
BHMT Monoclonal specifically detects BHMT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
BHMT | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BHMT | |
Full length recombinant protein of human BHMT (NP_001704) produced in HEK293T cells. | |
0.1 mL | |
635.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3E11 | |
Western Blot, Immunohistochemistry | |
betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction