Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHMT Monoclonal Antibody (OTI3E11)
GREENER_CHOICE

Catalog No. PIMA525054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525054 Supplier Invitrogen™ Supplier No. MA525054
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BHMT Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHMT
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3E11
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BHMT
Gene Accession No. Q93088
Gene Alias betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p
Gene Symbols BHMT
Host Species Mouse
Immunogen Full length human recombinant protein of BHMT produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 635
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.